Assay Details
Gene Symbol: LOC5570587 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001653202.3 | XP_001653252.2 | 2 - 3 | 266 | 68 | |
XM_021842776.1 | XP_021698468.1 | 2 - 3 | 397 | 68 | ||
GenBank mRNA | AF394234.1 | - | 2 - 3 | 217 | 68 | |
AY432888.1 | - | 2 - 3 | 231 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() |
