Assay Details
Gene Symbol: LOC5570587 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001653202.3 | XP_001653252.2 | 3 - 4 | 486 | 81 | |
XM_021842776.1 | XP_021698468.1 | 3 - 4 | 617 | 81 | ||
GenBank mRNA | AF394234.1 | - | 3 - 4 | 437 | 81 | |
AY432888.1 | - | 3 - 4 | 451 | 81 |

More Information
Set Membership: |
![]() ![]() ![]() |
