Assay Details
Gene Symbol: LOC5564787 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001649093.2 | XP_001649143.1 | 1 - 2 | 334 | 61 | |
GenBank mRNA | AF395204.1 | - | 1 - 2 | 312 | 61 |

More Information
Set Membership: |
![]() ![]() ![]() |
