Assay Details
Gene Symbol: LOC5564787 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001649093.2 | XP_001649143.1 | 3 - 4 | 1019 | 65 | |
GenBank mRNA | AF395204.1 | - | 3 - 4 | 997 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() |
