Assay Details
Gene Symbol: LOC5566826 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021842414.1 | XP_021698106.1 | 1 - 2 | 163 | 70 | |
GenBank mRNA | AF466610.1 | - | 1 - 2 | 142 | 70 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021842414.1 transcripts.
Set Membership: |
![]() ![]() |
