Assay Details
Gene Symbol: LOC5566826 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021842411.1 | XP_021698103.1 | 2 - 3 | 1526 | 109 | |
XM_021842412.1 | XP_021698104.1 | 3 - 4 | 1444 | 109 | ||
XM_021842413.1 | XP_021698105.1 | 3 - 4 | 1449 | 109 | ||
XM_021842414.1 | XP_021698106.1 | 2 - 3 | 1329 | 109 | ||
GenBank mRNA | AF466610.1 | - | 2 - 3 | 1308 | 109 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
