Assay Details
Gene Symbol: LOC5575886 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021854189.1 | XP_021709881.1 | 2 - 3 | 304 | 65 | |
XM_021854200.1 | XP_021709892.1 | 2 - 3 | 304 | 65 | ||
GenBank mRNA | AF474405.1 | - | 1 - 2 | 274 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
