Assay Details
Gene Symbol: LOC5574983 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001655406.2 | XP_001655456.2 | 2 - 3 | 1045 | 64 | |
GenBank mRNA | AY249245.1 | - | 1 - 2 | 546 | 64 |

More Information
Set Membership: |
![]() ![]() ![]() |
