Assay Details
Gene Symbol: LOC5575011 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021852568.1 | XP_021708260.1 | 2 - 3 | 141 | ||
XM_021852569.1 | XP_021708261.1 | 2 - 3 | 141 | |||
GenBank mRNA | AY326254.1 | - | 2 - 3 | 905 | 141 |

More Information
Set Membership: |
![]() ![]() ![]() |
