Assay Details
Gene Symbol: LOC5575011 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021852568.1 | XP_021708260.1 | 3 - 4 | 73 | ||
XM_021852569.1 | XP_021708261.1 | 3 - 4 | 73 | |||
GenBank mRNA | AY326254.1 | - | 3 - 4 | 1018 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() |
