Assay Details
Gene Symbol: LOC5575011 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_011494871.2 | XP_011493173.2 | 3 - 4 | 941 | 86 | |
XM_021852568.1 | XP_021708260.1 | 5 - 6 | 1235 | 86 | ||
XM_021852569.1 | XP_021708261.1 | 5 - 6 | 1372 | 86 | ||
GenBank mRNA | AY326254.1 | - | 5 - 6 | 1340 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
