Assay Details
Gene Symbol: LOC5570007 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001658846.2 | XP_001658896.1 | 3 | 1963 | 63 | |
GenBank mRNA | AY431140.1 | - | 1 | 720 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
