Assay Details
Gene Symbol: LOC5575179 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021846584.1 | XP_021702276.1 | 6 - 7 | 865 | 83 | |
GenBank mRNA | AY431160.1 | - | 2 - 3 | 403 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
