Assay Details
Gene Symbol: LOC5575683 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001655759.2 | XP_001655809.1 | 2 - 3 | 1319 | 70 | |
XM_001655760.2 | XP_001655810.1 | 2 - 3 | 1318 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
