Assay Details
Gene Symbol: LOC5574193 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001661200.3 | XP_001661250.1 | 1 - 2 | 365 | 66 | |
GenBank mRNA | AY432974.1 | - | 1 - 2 | 154 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
