Assay Details
Gene Symbol: LOC5574193 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001661200.3 | XP_001661250.1 | 3 - 4 | 688 | 104 | |
GenBank mRNA | AY432974.1 | - | 3 - 4 | 477 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() |
