Assay Details
Gene Symbol: LOC5574943 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001655359.2 | XP_001655409.1 | 1 - 2 | 100 | ||
GenBank mRNA | AY432989.1 | - | 1 - 2 | 404 | 100 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
