Assay Details
Gene Symbol: LOC5565878 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001650196.2 | XP_001650246.1 | 1 - 2 | 163 | 78 | |
GenBank mRNA | AY433012.1 | - | 1 - 2 | 128 | 78 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001650196.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
