Assay Details
Gene Symbol: LOC5571387 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001653589.2 | XP_001653639.1 | 3 - 4 | 1003 | 60 | |
XM_001653590.2 | XP_001653640.1 | 4 - 5 | 815 | 60 | ||
XM_021841717.1 | XP_021697409.1 | 4 - 5 | 935 | 60 | ||
GenBank mRNA | AY431316.1 | - | 4 - 5 | 474 | 60 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
