Assay Details
Gene Symbol: LOC5574782 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001655289.2 | XP_001655339.1 | 4 - 5 | 2262 | 104 | |
GenBank mRNA | AY431377.1 | - | 1 - 2 | 191 | 104 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
