Assay Details
Gene Symbol: LOC5567501 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001651746.2 | XP_001651796.1 | 2 - 3 | 599 | 63 | |
GenBank mRNA | AY431379.1 | - | 1 - 2 | 290 | 63 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
