Assay Details
Gene Symbol: LOC5568395 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001657982.2 | XP_001658032.1 | 1 | 402 | 76 | |
GenBank mRNA | AY431399.1 | - | 1 | 326 | 76 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
