Assay Details
Gene Symbol: LOC5569643 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001658617.2 | XP_001658667.1 | 1 | 117 | ||
GenBank mRNA | AY431433.1 | - | 1 | 418 | 117 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
