Assay Details
Gene Symbol: LOC5580315 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001662987.3 | XP_001663037.1 | 6 | 3021 | 101 | |
GenBank mRNA | AY431495.1 | - | 1 | 416 | 101 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
