Assay Details
Gene Symbol: LOC5569647 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001652859.2 | XP_001652909.2 | 3 | 516 | 86 | |
XM_021852986.1 | XP_021708678.1 | 2 | 471 | 86 | ||
XM_021852987.1 | XP_021708679.1 | 2 | 996 | 86 | ||
GenBank mRNA | AY431541.1 | - | 1 | 417 | 86 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
