Assay Details
Gene Symbol: LOC5570675 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_011494753.2 | XP_011493055.1 | 1 - 2 | 96 | 75 | |
GenBank mRNA | AY431545.1 | - | 1 - 2 | 48 | 75 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_011494753.2 transcripts.
Set Membership: |
![]() ![]() ![]() |
