Assay Details
Gene Symbol: LOC5570675 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001659218.2 | XP_001659268.1 | 2 - 3 | 133 | 65 | |
XM_011494753.2 | XP_011493055.1 | 2 - 3 | 189 | 65 | ||
GenBank mRNA | AY431545.1 | - | 2 - 3 | 141 | 65 | |
CR937384.1 | - | 2 - 3 | 108 | 65 | ||
DQ440232.1 | - | 1 - 2 | 90 | 65 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
