Assay Details
Gene Symbol: LOC5576993 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021843881.1 | XP_021699573.1 | 11 | 2532 | 88 | |
XM_021843882.1 | XP_021699574.1 | 10 | 2482 | 88 | ||
GenBank mRNA | AY431596.1 | - | 1 | 122 | 89 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
