Assay Details
Gene Symbol: LOC5567423 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001657353.2 | XP_001657403.1 | 1 - 2 | 261 | 83 | |
GenBank mRNA | AY431621.1 | - | 1 - 2 | 261 | 83 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
