Assay Details
Gene Symbol: LOC5573805 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021845622.1 | XP_021701314.1 | 5 - 6 | 2713 | 70 | |
XM_021845624.1 | XP_021701316.1 | 5 - 6 | 2715 | 70 |

More Information
Set Membership: |
![]() ![]() ![]() |
