Assay Details
Gene Symbol: LOC5573805 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021845622.1 | XP_021701314.1 | 4 - 5 | 2532 | 66 | |
XM_021845624.1 | XP_021701316.1 | 4 - 5 | 2534 | 66 |

More Information
Set Membership: |
![]() ![]() ![]() |
