Assay Details
Gene Symbol: LOC5573805 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021845622.1 | XP_021701314.1 | 1 - 2 | 390 | 99 | |
XM_021845624.1 | XP_021701316.1 | 1 - 2 | 392 | 99 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021845622.1, XM_021845624.1 transcripts.
Set Membership: |
![]() ![]() |
