Assay Details
Gene Symbol: LOC110677186 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021848072.1 | XP_021703764.1 | 2 - 3 | 2497 | 111 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC110677186 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021848072.1 | XP_021703764.1 | 2 - 3 | 2497 | 111 |
Set Membership: |
![]() ![]() ![]() ![]() ![]() |