Assay Details
Gene Symbol: LOC5579949 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001652188.2 | XP_001652238.2 | 3 - 4 | 1216 | 105 | |
XM_021854387.1 | XP_021710079.1 | 4 - 5 | 2222 | 105 | ||
XM_021854388.1 | XP_021710080.1 | 4 - 5 | 2185 | 105 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
