Assay Details
Gene Symbol: LOC5564793 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001649067.2 | XP_001649117.1 | 2 - 3 | 258 | 117 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001649067.2 transcripts.
Set Membership: |
![]() ![]() ![]() ![]() |
