Assay Details
Gene Symbol: LOC5574407 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001655064.2 | XP_001655114.1 | 2 - 3 | 1845 | 82 | |
XM_021854649.1 | XP_021710341.1 | 2 - 3 | 1868 | 82 | ||
XM_021854650.1 | XP_021710342.1 | 2 - 3 | 1864 | 82 |
