Assay Details
Gene Symbol: LOC5579456 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021839461.1 | XP_021695153.1 | 7 | 2569 | 119 | |
XM_021839462.1 | XP_021695154.1 | 7 | 2562 | 119 | ||
XM_021839463.1 | XP_021695155.1 | 8 | 2826 | 119 | ||
XM_021839464.1 | XP_021695156.1 | 8 | 2816 | 119 | ||
XM_021839465.1 | XP_021695157.1 | 7 | 2514 | 119 | ||
XM_021839466.1 | XP_021695158.1 | 8 | 2812 | 119 | ||
XM_021839467.1 | XP_021695159.1 | 7 | 3312 | 119 | ||
GenBank mRNA | AY433105.1 | - | 1 | 221 | 119 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
