Assay Details
Gene Symbol: LOC5578955 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_011495462.2 | XP_011493764.2 | 4 | 1666 | 74 | |
GenBank mRNA | AY433125.1 | - | 1 | 520 | 74 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
