Assay Details
Gene Symbol: LOC5565856 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001650147.2 | XP_001650197.1 | 1 - 2 | 67 | ||
GenBank mRNA | AY433195.1 | - | 1 - 2 | 165 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
