Assay Details
Gene Symbol: LOC5579843 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021854134.1 | XP_021709826.1 | 4 - 5 | 494 | 73 | |
XM_021854135.1 | XP_021709827.1 | 4 - 5 | 499 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
