Assay Details
Gene Symbol: LOC5579843 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021854134.1 | XP_021709826.1 | 1 - 2 | 163 | 76 | |
XM_021854135.1 | XP_021709827.1 | 1 - 2 | 164 | 76 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_021854134.1, XM_021854135.1 transcripts.
Set Membership: |
![]() ![]() ![]() |
