Assay Details
Gene Symbol: LOC5564492 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001648782.2 | XP_001648832.1 | 6 - 7 | 1891 | 84 | |
XM_021845379.1 | XP_021701071.1 | 6 - 7 | 1843 | 84 |

More Information
Set Membership: |
![]() ![]() ![]() |
