Assay Details
Gene Symbol: LOC5573854 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001654831.2 | XP_001654881.1 | 1 - 2 | 231 | 70 | |
XM_021845591.1 | XP_021701283.1 | 1 - 2 | 232 | 70 |

More Information
Additional Information:
This assay is in the 5'UTR region of the XM_001654831.2, XM_021845591.1 transcripts.
Set Membership: |
![]() ![]() |
