Assay Details
Gene Symbol: LOC5578667 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021849550.1 | XP_021705242.1 | 4 - 5 | 1186 | 68 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC5578667 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021849550.1 | XP_021705242.1 | 4 - 5 | 1186 | 68 |
Set Membership: |
![]() ![]() ![]() ![]() |