Assay Details
Gene Symbol: LOC5578667 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021849550.1 | XP_021705242.1 | 3 - 4 | 1088 | 71 |

More Information
Set Membership: |
![]() ![]() ![]() |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: LOC5578667 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021849550.1 | XP_021705242.1 | 3 - 4 | 1088 | 71 |
Set Membership: |
![]() ![]() ![]() |