Assay Details
Gene Symbol: LOC5570153 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001658905.2 | XP_001658955.1 | 1 - 2 | 238 | 67 | |
GenBank mRNA | AY433250.1 | - | 1 - 2 | 102 | 67 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
