Assay Details
Gene Symbol: LOC5566857 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001651234.2 | XP_001651284.1 | 2 - 3 | 765 | 85 | |
XM_021854745.1 | XP_021710437.1 | 2 - 3 | 1269 | 85 | ||
GenBank mRNA | AY433257.1 | - | 2 - 3 | 701 | 85 | |
DQ440311.1 | - | 1 - 2 | 581 | 85 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() |
