Assay Details
Gene Symbol: LOC5578555 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001663897.2 | XP_001663947.1 | 3 - 4 | 466 | 103 | |
XM_021851902.1 | XP_021707594.1 | 3 - 4 | 489 | 103 | ||
XM_021851903.1 | XP_021707595.1 | 3 - 4 | 490 | 103 | ||
XM_021851904.1 | XP_021707596.1 | 3 - 4 | 463 | 103 |

More Information
Set Membership: |
![]() ![]() |
