Assay Details
Gene Symbol: LOC5578555 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_001663897.2 | XP_001663947.1 | 6 - 7 | 1115 | 65 | |
XM_021851902.1 | XP_021707594.1 | 6 - 7 | 1138 | 65 | ||
XM_021851903.1 | XP_021707595.1 | 6 - 7 | 1139 | 65 | ||
XM_021851904.1 | XP_021707596.1 | 6 - 7 | 1112 | 65 |

More Information
Set Membership: |
![]() ![]() |
