Assay Details
Gene Symbol: LOC5570359 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Exon Boundary | Assay Location | IMAGE Clone ID | Amplicon Length | |
RefSeq | XM_021851603.1 | XP_021707295.1 | 2 - 3 | 570 | 73 | |
XM_021851604.1 | XP_021707296.1 | 2 - 3 | 567 | 73 |

More Information
Set Membership: |
![]() ![]() ![]() ![]() ![]() |
